Alfa Aesar™ Platinum Rhodium gauze, 80 mesh woven from 0.076mm (0.003 in.) dia. wire, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 12217GH

Quantity 100x100mm

 View more versions of this product

Catalog No. AA12217GH

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00198186
    Chemical Name or Material Platinum Rhodium gauze, Woven from 0.076mm (0.003 in.) dia. wire

    Specifications

    MDL Number MFCD00198186
    Mesh Size 80 Mesh
    Odor Odorless
    Avg. Mol. Wt. or Mol. Wt. Range Pt:Rh; 90:10 wt%
    Weight ≈0.39g/25x25mm
    Quantity 100x100mm
    TSCA TSCA
    Chemical Name or Material Platinum Rhodium gauze, Woven from 0.076mm (0.003 in.) dia. wire
    Assay 99.9%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit