Alfa Aesar™ Platinum Rhodium Palladium gauze, 80 mesh woven from 0.076mm (0.003 in.) dia. wire, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 12216FI

Quantity 50x50mm

 View more versions of this product

Catalog No. AA12216FI

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00011179
    Chemical Name or Material Platinum Rhodium Palladium gauze, Woven from 0.076mm (0.003 in.) dia. wire

    Specifications

    TSCA TSCA
    MDL Number MFCD00011179
    Mesh Size 80 Mesh
    Avg. Mol. Wt. or Mol. Wt. Range Pt:Rh:Pd; 90:5:5 wt%
    Weight ≈0.39g/25x25mm
    Quantity 50x50mm
    Chemical Name or Material Platinum Rhodium Palladium gauze, Woven from 0.076mm (0.003 in.) dia. wire
    Assay 99.9%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit