Platinum Ruthenium black, nominally Pt 50%, Ru 50% (Atomic wt%), Alfa Aesar™

Manufacturer: Alfa Aesar 4117103

Quantity 1g

 View more versions of this product

Catalog No. AA4117103

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Solid
    Molecular Formula Pt:Ru; 50:50
    MDL Number MFCD00798552
    Chemical Name or Material Platinum Ruthenium black, HiSPEC 6000

    Specifications

    Quantity 1g
    Color Black
    DOT Information DOT Class: 4.1, Packaging Group: II
    Physical Form Solid
    Odor Odorless
    Molecular Formula Pt:Ru; 50:50
    MDL Number MFCD00798552
    Health Hazard 1 H228: Flammable solid.
    Health Hazard 2 Keep container tightly closed and dry. Keep away from sources of ignition - No Smoking. Take precautionary measures against static discharges.
    Chemical Name or Material Platinum Ruthenium black, HiSPEC 6000

    Keep container tightly closed and dry. Keep away from sources of ignition - No Smoking. Take precautionary measures against static discharges.

    H228: Flammable solid.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit