Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum Ruthenium foil, 0.05mm (0.002 in.) thick, Alfa Aesar™

Manufacturer:  Alfa Aesar 42686GH

Quantity 100 x 100mm

 View more versions of this product

Catalog No. AA42686GH


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

Foil
Pt:Ru; 95.2:4.8
MFCD00798552
Platinum Ruthenium

Specifications

100 x 100mm
Silver-gary foil
Foil
MFCD00798552
Silver-gray
Odorless
Pt:Ru; 95.2:4.8
Platinum Ruthenium
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit