Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum on Sulfide Carbon, 5% Pt, Contains ca 50% Moisture, Acros Organics

Click to view available options
Quantity:
10g
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black,platin,sponge,platinum, metal,platinum, elemental,platine,platino,metal,platin german,iv ion
23939
CHEBI:33400
platinum
[Pt]
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black,platin,sponge,platinum, metal,platinum, elemental,platine,platino,metal,platin german,iv ion
23939
CHEBI:33400
platinum
[Pt]
Quantity 10g
Fieser 01,892; 04,393; 01,982; 01,890; 15,268
Loss on Drying 56% (typically)
Formula Weight 195.08
Physical Form Powder
Percent Purity 5% (Pt)
Particle Size Typical particle size distribution: D50: 23 Micron, <8 Micron: 13%, <16 Micron: 33%, <32 Micron: 63%, <64 Micron: 88%, <128 Micron: 99%
Color Black
Chemical Name or Material Platinum on sulfide carbon