Invitrogen™ Platinum™ SuperFi™ DNA Polymerase

Amplify even difficult and GC-rich amplicons with superior accuracy (>100X Taq) and minimaloptimization steps

Manufacturer: LIFE TECHNOLOGIES 12351010

 View more versions of this product

Catalog No. 12-351-010

  Check Availability
Add to Cart

    Specifications

    For Use With (Application)
    • High-fidelity PCR
    • Cloning and sub-cloning
    • Site-directed mutagenesis
    • Amplification of GC-rich templates 
    • Template generation for sequencing
    • High-throughput PCR
    • Amplification of samples with suboptimal purity
    • Long PCR (up to 20 kb)
    • Fast PCR
    Includes Platinum SuperFi DNA Polymerase, 5X SuperFi Buffer, 5X SuperFi GC Enhancer
    Overhang Blunt
    Quantity 100 Units
    Format Separate components
    Storage Requirements Store at -20C.

    Invitrogen™ Platinum™ SuperFi™ DNA Polymerase is a proofreading DNA polymerase which combines superior fidelity with trusted PlatinumTM hot-start technology for the highest success in PCR. Featuring >100x Taq fidelity, Platinum SuperFi DNA Polymerase is ideally suited for cloning, mutagenesis and other applications benefiting from supreme sequence accuracy.

    • Receive a complimentary sample of Invitrogen™ Platinum™ SuperFi™ DNA Polymerase when you fill out the form on this page. Invitrogen Platinum SuperFi DNA Polymerase provides proofreading capability and adds superior fidelity to the trusted Platinum hot-start technology for PCR success.

      Buy Now

      Expires 2017-10-31

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit