Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Invitrogen™ Platinum™ Taq Green Hot Start DNA Polymerase Non-distribution product as customer accommodation.
Invitrogen™ Platinum™ Taq DNA Polymerase is a convenient and reliable “hot start” thermostable DNA polymerase for PCR that provides enhanced specificity over that of Taq DNA Polymerase.
Manufacturer:  Invitrogen™ 11966026
 View more versions of this product
Catalog No. 11-966-026
Description
Description
Invitrogen™ Platinum™ Taq DNA Polymerase is a convenient and reliable “hot start” thermostable DNA polymerase for PCR that provides enhanced specificity over that of Taq DNA Polymerase.
Specifications
Specifications
Store at -20°C in a non-frost-free freezer.
Each box contains: 60 µL Platinum Taq DNA Polymerase; 2 x 1.25 mL 10X Green PCR Buffer, -Mg; 1.25 mL 50 mM magnesium chloride; 1.25 mL KB Extender
3'-A
300 reactions
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.