Alfa Aesar™ Platinum wire, 0.0762mm (0.003 in.) dia., For ISA Type R or S Standard Grade Thermocouple

For ISA Type R or S Standard Grade Thermocouple

Manufacturer: Alfa Aesar™ 10291G2

Quantity 2m

 View more versions of this product

Catalog No. AA10291G2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-06-4
    MDL Number MFCD00011179
    Chemical Name or Material Platinum wire, 0.0762mm (0.003 in.) dia.

    Specifications

    TSCA TSCA
    Physical Form Wire
    MDL Number MFCD00011179
    CAS 7440-06-4
    Odor Odorless
    Weight ≈0.05g/50cm
    Quantity 2m
    Chemical Name or Material Platinum wire, 0.0762mm (0.003 in.) dia.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit