Platinum Wire, 0.076mm (0.003 in.) dia., Alfa Aesar™

Form: Approx .0.045g/50cm

Manufacturer: Alfa Aesar 10079BW

Quantity 50cm

 View more versions of this product

Catalog No. AA10079BW

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Wire, 0.076mm (0.003 in.) dia.
    Molecular Formula Pt:Rh; 70:30 wt%
    MDL Number MFCD00198186
    Chemical Name or Material Platinum, 30 wt% Rhodium wire

    Specifications

    Quantity 50cm
    Color Silver
    Physical Form Wire, 0.076mm (0.003 in.) dia.
    Molecular Formula Pt:Rh; 70:30 wt%
    MDL Number MFCD00198186
    Odor Odorless
    Appearance Silver colored wire
    Chemical Name or Material Platinum, 30 wt% Rhodium wire

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit