Platinum wire, 99.95% (metals basis), Alfa Aesar™

Form: approx. 16.8g/m; Temper: as drawn

Manufacturer: Alfa Aesar 10285BQ

Quantity 5cm

 View more versions of this product

Catalog No. AA10285BQ

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7440-06-4
    Physical Form Wire, 1.0mm (0.04 in.) dia.
    Assay Percent Range 99.95% (Metals basis)
    MDL Number MFCD00011179
    Chemical Name or Material Platinum, Hard

    Specifications

    Quantity 5cm
    Color Silvery white
    EINECS Number 231-116-1
    Physical Form Wire, 1.0mm (0.04 in.) dia.
    Assay Percent Range 99.95% (Metals basis)
    MDL Number MFCD00011179
    CAS 7440-06-4
    Odor Odorless
    Appearance Silvery white wire
    Chemical Name or Material Platinum, Hard

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit