Platinum wire, 0.25mm dia., Alfa Aesar™

Manufacturer: Alfa Aesar 10288CC

Quantity 500cm

 View more versions of this product

Catalog No. AA10288CC

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7440-06-4
    MDL Number MFCD00011179
    Chemical Name or Material Platinum
    Grade For ISA Type R or S Standard Grade

    Specifications

    Quantity 500cm
    Density 21.45g/cm3 at 20°C
    EINECS Number 231-116-1
    Physical Form Wire, 0.25mm (0.01 in.) dia.
    MDL Number MFCD00011179
    CAS 7440-06-4
    Odor Odorless
    Appearance Silvery white wire
    Chemical Name or Material Platinum
    Grade For ISA Type R or S Standard Grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit