Learn More
PLAU Rabbit anti-Human, Unconjugated, Clone: AABC-16, Abnova™
Rabbit Monoclonal Antibody.
Manufacturer:  Abnova Corporation MAB20833
Catalog No. 89-337-233
Add to cart
Description
Description
PLAU Monoclonal antibody specifically detects PLAU in Human samples. It is validated for IHC, WB-Ce.
Specifications
Specifications
PLAU
AABC-16
plasminogen activator, urokinase
Liquid
Rabbit
IgG
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
PLAU
ATF, UPA, URK, UROKINASE, u-PA
A synthetic peptide corresponding to human PLAU
100 μg
Primary
5328