Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLC-delta 3 Goat anti-Human, Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Supplier: R&D Systems AF5315SP
This item is not returnable.
View return policy
Description
PLC-delta 3 Polyclonal specifically detects PLC-delta 3 in Human, Mouse samples. It is validated for Western Blot.Specifications
PLC-delta 3 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.1.4.11, KIAA1964, MGC71172, Phosphoinositide phospholipase C-delta-3, phospholipase C, delta 3,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-3, Phospholipase C-delta-3, PLC delta3, PLCD3, PLCdelta 3, PLC-delta-3 | |
Goat | |
Affinity Purified | |
RUO | |
113026 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q8N3E9 | |
PLCD3 | |
E. coli-derived recombinant human PLC-delta 3 Lys148-Ala344 Accession # Q8N3E9 | |
25 μg | |
Primary | |
Detects human and mouse PLC-delta 3 in Western blots. | |
Human, Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only