Learn More
PLC-gamma 1 (p Tyr783) Mouse anti-Human, DyLight 488, Clone: MM0870-47Q33, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236791G
Catalog No. NBP236791G
Add to cart
Description
Description
PLC-gamma 1 p Tyr783 Monoclonal antibody specifically detects PLC-gamma 1 p Tyr783 in Human samples. It is validated for Western Blot.
Specifications
Specifications
PLC-gamma 1 (p Tyr783)
MM0870-47Q33
Western Blot
Purified
PLCG1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells) from a mouse immunized with Phosphopeptide containing Y783 of human PLC gamma -1.
Protein G purified
Lipid and Metabolism, Phospho Specific, Signal Transduction
Primary
5335
Human
Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
EC 3.1.4.11, gamma 1 (formerly subtype 148), monophosphatidylinositol phosphodiesterase, NCKAP3,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-1, phosphatidylinositol phospholipase C, phosphoinositidase C, phosphoinositide phospholipase C, Phosphoinositide phospholipase C-gamma-1, phospholipase C, gamma 1, phospholipase C-148, Phospholipase C-gamma-1, Phospholipase C-II, PLC11-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma 1, PLC148, PLC-148, PLC-gamma-1, PLC-II1-phosphatidyl-D-myo-inositol-4,5-bisphosphate, triphosphoinositide phosphodiesterase
Mouse
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody recognizes human PLC gamma-1 of pY783 in direct ELISAs and WBs.