Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLC-gamma 2 Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0873-19S8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236793AF350

Catalog No. NP236793A30


Add to cart

Description

Description

PLC-gamma 2 Monoclonal antibody specifically detects PLC-gamma 2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

PLC-gamma 2
MM0873-19S8
Western Blot
Conjugated, Purified
PLCG2
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells) from a mouse immunized with Phosphopeptide containing Y753 of human PLC gamma -2.
Protein G purified
Lipid and Metabolism, Phospho Specific
Primary
5336.0
Human
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
EC 3.1.4.11, Phosphoinositide phospholipase C-gamma-2, phospholipase C gamma 2, phospholipase C, gamma 2 (phosphatidylinositol-specific), phospholipase C, gamma 2 (phosphatidylyinositol-specific), Phospholipase C-gamma-2,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-2, Phospholipase C-IV, PLCG2, PLC-gamma-2, PLC-IV
Mouse
IgG2
0.1 mL
Store at 4°C in the dark.
Monoclonal
This antibody recognizes human PLC gamma-2 of pY753 in direct ELISAs and WBs.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit