Learn More
PLC-gamma 2 (p Tyr753) Mouse anti-Human, Biotin, Clone: MM0873-19S8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236793B
Catalog No. NBP236793B
Add to cart
Description
Description
PLC-gamma 2 p Tyr753 Monoclonal antibody specifically detects PLC-gamma 2 p Tyr753 in Human samples. It is validated for Western Blot.
Specifications
Specifications
PLC-gamma 2 (p Tyr753)
MM0873-19S8
Western Blot
Purified
PLCG2
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells) from a mouse immunized with Phosphopeptide containing Y753 of human PLC gamma -2.
Protein G purified
Lipid and Metabolism, Phospho Specific
Primary
5336
Human
Western Blot
Biotin
PBS with 0.05% Sodium Azide
EC 3.1.4.11, Phosphoinositide phospholipase C-gamma-2, phospholipase C gamma 2, phospholipase C, gamma 2 (phosphatidylinositol-specific), phospholipase C, gamma 2 (phosphatidylyinositol-specific), Phospholipase C-gamma-2,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-2, Phospholipase C-IV, PLCG2, PLC-gamma-2, PLC-IV
Mouse
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody recognizes human PLC gamma-2 of pY753 in direct ELISAs and WBs.