Learn More
PLC-gamma 2 (p Tyr759) Mouse anti-Human, FITC, Clone: MM0872-27K18, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236792F
Catalog No. NBP236792F
Add to cart
Description
Description
PLC-gamma 2 p Tyr759 Monoclonal antibody specifically detects PLC-gamma 2 p Tyr759 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
PLC-gamma 2 (p Tyr759)
MM0872-27K18
Immunocytochemistry/Immunofluorescence
Purified
PLCG2
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells) from a mouse immunized with Phosphopeptide containing Y759 of human PLC gamma -2
Protein G purified
Lipid and Metabolism, Phospho Specific
Primary
5336
Human
Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
EC 3.1.4.11, Phosphoinositide phospholipase C-gamma-2, phospholipase C gamma 2, phospholipase C, gamma 2 (phosphatidylinositol-specific), phospholipase C, gamma 2 (phosphatidylyinositol-specific), Phospholipase C-gamma-2,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-2, Phospholipase C-IV, PLCG2, PLC-gamma-2, PLC-IV
Mouse
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody recognizes human PLC gamma-2 of pY759 in direct ELISAs and WBs.