Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLCH2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103474
Description
Antibody detects endogenous levels of total PLCH2.
PLCH2 is a member of the PLC-eta family of the phosphoinositide-specific phospholipase C (PLC) superfamily of enzymes that cleave PtdIns(4,5) P2 to generate second messengers inositol 1,4,5-trisphosphate and diacylglycerol (Zhou et al., 2005 [PubMed 16107206]).
Specifications
PLCH2 | |
Polyclonal | |
Unconjugated | |
PLCH2 | |
1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase eta-2; A930027K05Rik; KIAA0450; Phosphoinositide phospholipase C-eta-2; phosphoinositide phospholipase C-like 4; phospholipase C eta 2; phospholipase C, eta 2; phospholipase C-eta2; Phospholipase C-eta-2; phospholipase C-like 4; phospholipase C-like protein 4; PLCeta2; Plc-eta2; PLCH2; PLCL4; PLC-L4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
269615, 9651 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
A2AP18, O75038 | |
PLCH2 | |
A synthesized peptide derived from human PLCH2(Accession O75038), corresponding to amino acid residues V142-G192. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction