Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLCXD1 Mouse anti-Human, DyLight™ 755, Clone: OTI2D7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273458IR
Description
PLCXD1 Monoclonal specifically detects PLCXD1 in Human samples. It is validated for Immunohistochemistry.Specifications
PLCXD1 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PLCXD1 | |
Full length recombinant protein of human PLCXD1 (NP_060860) produced in E.coli. | |
0.1 mL | |
55344.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI2D7 | |
Immunohistochemistry | |
FLJ11323, phosphatidylinositol-specific phospholipase C, X domain containing 1, PI-PLC X domain-containing protein 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title