Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLD2, Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

$119.60 - $398.00

Specifications

Antigen PLD2
Applications Western Blot, Immunohistochemistry
Host Species Goat
Target Species Human
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AF10123100
View Documents
R&D Systems
AF10123100
100 ug
Each for $398.00
Add to cart
 
AF10123SP
View Documents
R&D Systems
AF10123SP
25 ug
Each for $119.60
Add to cart
 
Description

Description

PLD2 Polyclonal antibody specifically detects PLD2 in Human Samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

PLD2
Goat
Polyclonal
Western Blot, Immunohistochemistry
Human
Primary
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit