Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pleckstrin-2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30339520UL
Description
Pleckstrin-2 Polyclonal specifically detects Pleckstrin-2 in Mouse samples. It is validated for Western Blot.Specifications
Pleckstrin-2 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Unconjugated | |
pleckstrin 2, pleckstrin 2 homolog, pleckstrin-2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human Pleckstrin-2 (NP_057529.1). | |
20 μL | |
Signal Transduction | |
26499 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction