Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pleckstrin-2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$183.00 - $490.00
Specifications
| Antigen | Pleckstrin-2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
Pleckstrin-2 Polyclonal specifically detects Pleckstrin-2 in Mouse samples. It is validated for Western Blot.Specifications
| Pleckstrin-2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| pleckstrin 2, pleckstrin 2 homolog, pleckstrin-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human Pleckstrin-2 (NP_057529.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 26499 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title