Learn More
Pleckstrin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA592885
Description
Immunogen sequence: MEPKRIREGY LVKKGSVFNT WKPMWVVLLE DGIEFYKKKS DNSPKGMIPL KGSTLTSPCQ DFGKRMFVFK ITTTKQQDHF FQAAFLEERD AWVRDIKKAI KCIEGGQKFA RKSTRRSIRL PETIDLGALY LSMKDTEKGI KELNLEKDKK IFNHCFTGNC VIDWLVSNQS VRNRQEGLMI ASSLLNEGYL QPAGDMSKSA VDGTAENPFL DNPDAFYYFP DSGFFCEENS SDDDVILKEE FRGVIIKQGC LLKQGHRRKN WKVRKFILRE DPAYLHYYDP AGAEDPLGAI HLRGCVVTSV ESNSNGRKSE EENLFEIITA DEVHYFLQAA TPKERTEWIR AIQMASRTGK; Positive Samples: THP-1, U-937, Mouse lung, Mouse spleen, Rat spleen
Major protein kinase C substrate of platelets.Specifications
Pleckstrin | |
Polyclonal | |
Unconjugated | |
PLEK | |
PLEK | |
Recombit fusion protein containing a sequence corresponding to amino acids 1-350 of human PLEK | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
2.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08567, Q4KM33, Q9JHK5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
364206, 5341, 56193 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.