Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pleiotrophin Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550567

Catalog No. PA550567


Add to cart

Description

Description

The antibody detects endogenous levels of total Pleiotrophin protein.

Pleiotrophin (PTN) is an 18 kDa secreted heparin and chondroitin-binding protein that is a regulatory cytokine critical to the early development of blood vessels and neurons. High PTN serum levels are associated with a variety of solid tumors. It was shown that patients with multiple myeloma (MM) also have elevated serum levels of this protein. The expression of this protein is very limited in normal adult tissues.
Specifications

Specifications

Pleiotrophin
Polyclonal
Unconjugated
PTN
Liquid
PTN
Synthetic peptide corresponding to a region derived from internal residues of human pleiotrophin
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1.2 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
P21246, P63089, P63090
HARP, HBGF8, HBNF, NEGF1
Rabbit
IgG
100 μL
-20°C
Primary
19242, 24924, 5764
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit