Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pleiotrophin Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551122

Catalog No. PA551122


Add to cart

Description

Description

Pleiotrophin Polyclonal Antibody detects endogenous levels of Pleiotrophin protein.

Secreted growth factor that induces neurite outgrowth and which is mitogenic for fibroblasts, epithelial, and endothelial cells. Binds anaplastic lymphoma kinase (ALK) which induces MAPK pathway activation, an important step in the anti-apoptotic signaling of PTN and regulation of cell proliferation. {ECO:0000269; PubMed:11278720, ECO:0000269; PubMed:1768439}.
Specifications

Specifications

Pleiotrophin
Polyclonal
Unconjugated
PTN
Liquid
PTN
Synthesized peptide derived from the Internal region of human PTN.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P21246, P63089, P63090
HARP, HBGF8, HBNF, NEGF1
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
19242, 24924, 5764
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit