Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pleiotrophin/PTN Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$366.50 - $607.50
Specifications
Antigen | Pleiotrophin/PTN |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Pleiotrophin/PTN Polyclonal antibody specifically detects Pleiotrophin/PTN in Human samples. It is validated for ImmunofluorescenceSpecifications
Pleiotrophin/PTN | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Cell Cycle and Replication | |
PBS, pH 7.2, 40% glycerol | |
5764 | |
IgG | |
Affinity purified |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
Polyclonal | |
Purified | |
RUO | |
Human | |
HARP, HBBM, HB-GAM, HBGF8, HBNF, HBNF1, HBNF-1, Heparin-binding brain mitogen, NEGF1HBGF-8, neurite growth-promoting factor 1, neurite growth-promoting factor1), OSF-1, Osteoblast-specific factor 1, pleiotrophin | |
This antibody was developed against Recombinant Protein corresponding to amino acids: KQFGAECKYQFQAWGECDLNTALKTRTGSLKRALHNAECQKTVTISKPCGKLTKPKPQAESKKK | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title