Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLEKHA4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548272
Description
Binds specifically to phosphatidylinositol 3-phosphate (PtdIns3P), but not to other phosphoinositides.Specifications
PLEKHA4 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9H4M7 | |
PLEKHA4 | |
KLH conjugated synthetic peptide between 21-50 amino acids from the N-terminal region of human PLEKHA4. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
PLEKHA4 | |
Pleckstrin homology domain-containing family A member 4, PH domain-containing family A member 4, Phosphoinositol 3-phosphate-binding protein 1, PEPP-1, PLEKHA4, PEPP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57664 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction