Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLEKHA7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584708
Description
Immunogen sequence: ALKENKDQLE SVLEVLHRQM EQYRDQPQHL EKIAYQQKLL QEDLVHIRAE LSRESTEMEN AWNEYLKLEN DVEQLKQTLQ EQHRRAFFFQ EKSQIQKDLW RIEDVTA.
PLEKHA7 is required for zonula adherens biogenesis and maintenance. Acts via its interaction with KIAA1543/Nezha, which anchors microtubules at their minus-ends to zonula adherens, leading to the recruitment of KIFC3 kinesin to the junctional site.
Specifications
PLEKHA7 | |
Polyclonal | |
Unconjugated | |
PLEKHA7 | |
A430081P20Rik; AI414265; DKFZp686M22243; Hadp1; Heart adapter protein 1; PH domain-containing family A member 7; PH domain-containing family A member 7 antibody; pleckstrin homology domain containing A7; pleckstrin homology domain containing A7 antibody; pleckstrin homology domain containing, family A member 2; pleckstrin homology domain containing, family A member 7; pleckstrin homology domain-containing family A member 7; Plekha7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
144100 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6IQ23 | |
PLEKHA7 | |
Recombinant protein corresponding to Human PLEKHA7. Recombinant protein control fragment (Product #RP-108512). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction