Learn More
Aviva Systems Biology Corp PLEKHA9 Antibody - N-terminal region (ARP56767_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56767_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLEKHA9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PLEKHA9. Alternate names: FLJ14156, PLEKHA9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61981