Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLEKHA9 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | PLEKHA9 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP170675
|
Novus Biologicals
NBP170675 |
100 μL |
Each of 1 for $436.00
|
|
Description
PLEKHA9 Polyclonal specifically detects PLEKHA9 in Human samples. It is validated for Western Blot.Specifications
PLEKHA9 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
51054 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
PLEKHA8P1 pleckstrin homology domain containing, family A member 8 pseudogene 1 | |
PLEKHA9 | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title