Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLEKHO1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520827
Description
A suggested positive control is human lung tissue lysate. PA5-20827 can be used with blocking peptide PEP-0941.
CKIP1 was identified through a yeast-two hybrid screening as a protein that would interact with only one of the two catalytic subunits of the casein kinase 2 complex. CKIP1 is a pleckstrin homology domain-containing protein localized within the nucleus and at the plasma membrane that interacts with CK2alpha but not CK2alpha', and is thought to play a role in targeting CK2alpha to a particular cellular location. CKIP1 has been implicated in muscle differentiation and the regulation of cell morphology and actin cytoskeleton. CKIP1 can also interact with other proteins such as ATM, an upstream kinase of p53, and recruit the nuclear ATM to the plasma membrane, suggesting CKIP1 may regulate ATM function through re-localizing ATM. CKIP1 can also form a complex with the kinase AKT, leading to a down-regulation of PI3K/AKT signaling and suppression of tumor growth in vivo.
Specifications
PLEKHO1 | |
Polyclonal | |
Unconjugated | |
PLEKHO1 | |
2810052M02Rik; Casein kinase 2-interacting protein 1; C-Jun-binding protein; CK2 interacting protein 1; CK2 interacting protein 1; HQ0024c protein; CK2-interacting protein 1; CKIP1; CKIP-1; HQ0024c; HQ0024c protein; JBP; Jun zipper-associated 2; Jza2; JZA-20; OC120; Osteoclast maturation-associated gene 120 protein; PH domain-containing family O member 1; pleckstrin homology domain containing O1; pleckstrin homology domain containing, family O member 1; pleckstrin homology domain-containing family O member 1; pleckstrin homology domain-containing family O member 1-like; PLEKHO1; RGD1311487; RP11-458I7.3; TNF intracellular domain-interacting protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
310674, 51177, 67220 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q53GL0, Q5BJM5, Q9JIY0 | |
PLEKHO1 | |
An 18 amino acid peptide from near the carboxy terminus of human CKIP1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction