Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plexin-B2 Armenian Hamster anti-Mouse, Clone: eBio3E7 (3E7), eBioscience™ Non-distribution product as customer accommodation.

Armenian Hamster Monoclonal Antibody

Manufacturer:  Invitrogen 14566581

 View more versions of this product

Catalog No. 50-131-13


Add to cart

Description

Description

Description: Plexin-B2 is a member of the Semaphorin receptor family. The semaphorin family consists of membrane and secreted proteins involved in attraction and repulsion of neuronal guidance cues. Plexin-B1 is a transmembrane protein that can be cleaved coverting the single chain presursor into a heterodimeric receptor. It has been shown to bind Sema4D/CD100 and is involved in axon development in mouse embryos. Sema4D also plays a role in the immune system interacting with CD72. Plexin-B2 ft.s possible role in the immune system has not been characterized. Applications Reported: This eBio3E7 (3E7) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistochemical staining. Applications Tested: This eBio3E7 (3E7) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered.

Plexin-B2 is a member of the Semaphorin receptor family. The semaphorin family consists of membrane and secreted proteins involved in attraction and repulsion of neuronal guidance cues. Plexin-B1 is a transmembrane protein that can be cleaved coverting the single chain presursor into a heterodimeric receptor. It has been shown to bind Sema4D/CD100 and is involved in axon development in mouse embryos. Sema4D also plays a role in the immune system interacting with CD72. Plexin-B2 ft.s possible role in the immune system has not been characterized.
Specifications

Specifications

Plexin-B2
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
B2RXS4
MM1, PLEXB2, PLXNB2, MM1KIAA0315dJ402G11.3, Nbla00445, PLEXB2, plexin B2, plexin-B2
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry, Immunohistochemistry, Immunoprecipitation
eBio3E7 (3E7)
Unconjugated
Plxnb2
Liquid
Plxnb2
IgG
50 μg
4° C
Primary
140570
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.