Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plexin C1 (CD232) Mouse, Alexa Fluor 647, Clone: 544232, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 566249

Catalog No. BDB566249


Add to cart

Description

Description

The 544232 monoclonal antibody specifically recognizes human and mouse Plexin C1 (PLXNC1) which is also known as, Virus-encoded semaphorin protein receptor (VESPR), or CD232. Plexin C1 is a ñ210 kDa single-pass type I membrane glycoprotein that belongs to the C subgroup of the plexin family of proteins. It is expressed on monocytes, dendritic cells, and neutrophils, as well as, some B lymphocytes and neuronal cells. Plexin C1 binds to Semaphorin 7A (CD108) and the viral semaphorin-like protein, A39R. Plexin C1 reportedly plays a role in cellular adhesion, migration, and phagocytosis.

Alexa Fluor™ 647 conjugates are highly photostable and remain fluorescent over a broad pH range. The excitation and emission maxima are nearly identical to those of APC. However, APC tends to be brighter while Alexa Fluor™ 647 is more optimal for intracellular applications. This fluorochrome exhibits uncommon photostability, making it an ideal choice for use in fluorescence microscopy. Due to nearly identical excitation and emission properties but different spillover characteristics, APC and Alexa Fluor™ 647 cannot be used simultaneously.
Specifications

Specifications

Plexin C1 (CD232)
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
CD232, Plexin-C1, plexinC1, PLXC1, PLXN-C1, PLXNC1, VESPR
Human Plexin C1 Recombinant Protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions, and unreacted Alexa Fluor™ 647 was removed.
RUO
Primary
Bovine, Human
544232
Alexa Fluor 647
Affinity Purified
Murine
Mouse IgG2b, κ
25μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit