Learn More
PlGF-1 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
Supplier: Pepro Tech Inc 500P226BT25UG

Description
AA Sequence of recombinant protein: MLPAVPPQQW ALSAGNGSSE VEVVPFQEVW GRSYCRALER LVDVVSEYPS EVEHMFSPSC VSLLRCTGCC GDENLHCVPV ETANVTMQLL KIRSGDRPSY VELTFSQHVR CECRPLREKM KPERCGDAVP RR. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human PlGF-1. Anti-Human PlGF-1-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hPlGF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human PlGF (500-P226) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hPlGF. Western Blot: To detect hPlGF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hPlGF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
PlGF-1 Polyclonal antibody specifically detects PlGF-1 in Human samples. It is validated for ELISA, Western BlotSpecifications
PlGF-1 | |
Polyclonal | |
PBS with no preservative | |
P49763 | |
E.coli-derived, 29.7 kDa Recombinant Human PlGF-1 | |
25 μg | |
Primary | |
Human | |
Lyophilized |
ELISA, Western Blot | |
Biotin | |
PGF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5228 | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.