Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLGRKT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552532
Description
Immunogen sequence: QKEFMLMNAR LQLERQLIMQ SEMRERQMAM QIAWSRE Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 89%.
The exact function of C9orf46 remains unknown.
Specifications
PLGRKT | |
Polyclonal | |
Unconjugated | |
PLGRKT | |
1110007H22Rik; 5033414D02Rik; AD025; AI852040; C9orf46; MDS030; Plasminogen receptor (KT); plasminogen receptor with a C-terminal lysine; plasminogen receptor, C-terminal lysine transmembrane protein; Plg-R(KT); PLGRKT; Plg-RKT; RGD1306839; transmembrane protein C9orf46; transmembrane protein C9orf46 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55848 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HBL7 | |
PLGRKT | |
Recombinant protein corresponding to Human PLGRKT. Recombinant protein control fragment (Product #RP-90083). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction