Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLK1 Antibody (AZ24), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002792JF549
Description
PLK1 Monoclonal antibody specifically detects PLK1 in Human, Mouse, Xenopus samples. It is validated for Western Blot, ELISASpecifications
PLK1 | |
Monoclonal | |
Janelia Fluor 549 | |
cell cycle regulated protein kinase, EC 2.7.11, EC 2.7.11.21, PLK, PLK-1, polo (Drosophia)-like kinase, polo like kinase, polo-like kinase 1polo-like kinase (Drosophila), Serine/threonine-protein kinase 13, serine/threonine-protein kinase PLK1, STPK13 | |
Baculo expressed Polo Kinase from Xenopus laevis | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Stem Cell Markers | |
5347 | |
Store at 4°C in the dark. | |
IgG2a |
Western Blot, ELISA | |
AZ24 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Xenopus species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction