Learn More
PLK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 159561AP
Catalog No. 50-173-0598
Add to cart
Description
Description
Serum-inducible kinase is a member of the  ft.polo ft. family of serine/threonine protein kinases that have a role in normal cell division.
Specifications
Specifications
PLK2
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P53351, Q9NYY3, Q9R012
PLK 2, PLK2, Polo like kinase 2, Serum inducible kinase, SNK
Rabbit
IgG
150 μL
-20°C
Primary
10769, 20620, 83722
Immunoprecipitation, Western Blot
Unconjugated
PLK2
Liquid
PLK2
PLK2 Fusion Protein Ag8710
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat