Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLK3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548707

Catalog No. PA548707

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CNK, a member of the CDC5/Polo subfamily of Ser/Thr protein kinases, is involved in regulating M phase functions during the cell cycle. It may also be part of the signaling network controlling cellular adhesion. In vitro, is able to phosphorylate CDC25C and casein. This membrane-associated protein binds to the calcium/integrin-binding protein (CIB). This interaction probably occurs via the POLO-box domain. Transcripts are highly detected in placenta, lung, followed by skeletal muscle, heart, pancreas, ovaries and kidney and weakly detected in liver and brain. This protein exhibits a short half-live. In cells of hematopoietic origin, CNK is strongly and exclusively detected in terminally differentiated macrophages. Transcript expression appears to be down-regulated in primary lung tumor. Cytokine and cellular adhesion trigger CNK induction. CNK is thought to be phosphorylated as cells enter mitosis and dephosphorylated as cells exit mitosis The protein contains 2 POLO box domains.
Specifications

Specifications

Plk3
Polyclonal
PBS with 0.09% sodium azide
Q9H4B4, Q60806
Serine/threonine-protein kinase PLK3, Cytokine-inducible serine/threonine-protein kinase, FGF-inducible kinase, Polo-like kinase 3, PLK-3, Plk3, Cnk, Fnk
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1263, 12795
Flow Cytometry, Western Blot
Unconjugated
PLK3
Liquid
PLK3
KLH conjugated synthetic peptide between 33-61 amino acids from the N-terminal region of mouse Plk3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit