Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLK5 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548678
Description
Inactive serine/threonine-protein kinase that plays a role in cell cycle progression and neuronal differentiation.
Specifications
Plk5 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q4FZD7 | |
PLK5 | |
KLH conjugated synthetic peptide between 4-31 amino acids from the N-terminal region of mouse Plk5. | |
400 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
Plk5 | |
Inactive serine/threonine-protein kinase PLK5, Polo-like kinase 5, PLK-5, Plk5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
216166 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction