Learn More
Invitrogen™ PLLP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120398
Description
Positive test controls include: Mouse brain, Rat brain, Rat kidney. The target is usually found in the following locations: Membrane, Multi-pass membrane protein. Immunogen sequence: MAEFPSKVST RTSSPAQGAE ASVSALRPDL GFVRSRLGAL MLLQLVLGLL VWALIADTPY HLYPAYGWVM FVAVFLWLVT IVLFNLYLFQ LHMKLYMVPW.
Plasmolipin is an 18 kDa membrane-bound proteolipid that belongs to the tetraspan (4TM) family, a rapidly growing group of myelin-associated proteins. Many of these proteins could be linked to human hereditary demyelinating neuropathies. Studies indicate that plasmolipin is a component of the cytoskeletal structure and has been localized to the apical surface of tubular epithelial cells. It has been shown that, upon addition to lipid bilayers, plasmolipin induces formation of ion channels that are both voltage-dependent and potassium-specific. Plasmolipin was originally isolated from kidney tissue. Subsequently, it has been shown to be widely expressed, not only being present in kidney, but also in the nervous system, thymus, testis, lung, and thyroid gland.
Specifications
PLLP | |
Polyclonal | |
Unconjugated | |
PLLP | |
0610010I06Rik; AV001002; Plapi; Plasma membrane proteolipid; plasma membrane proteolipid (plasmolipin); plasmolipin; PLLP; PMLP; Tm4sf11; transmembrane 4 superfamily member 11; transmembrane 4 superfamily member 11 (plasmolipin) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
64364, 67801 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.15 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P47987, Q9DCU2 | |
PLLP | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PLLP (NP_057077.1). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.