Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLOD1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548414

Catalog No. PA548414


Add to cart

Description

Description

Predicted to react with mouse based on sequence homology.

Forms hydroxylysine residues in -Xaa-Lys-Gly- sequences in collagens. These hydroxylysines serve as sites of attachment for carbohydrate units and are essential for the stability of the intermolecular collagen cross-links.
Specifications

Specifications

PLOD1
Polyclonal
PBS with 0.09% sodium azide
Q02809
Procollagen-lysine, 2-oxoglutarate 5-dioxygenase 1, Lysyl hydroxylase 1, LH1, PLOD1, LLH, PLOD
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5351
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PLOD1
Liquid
PLOD1
KLH conjugated synthetic peptide between 66-94 amino acids from the N-terminal region of human PLOD1.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit