Learn More
Aviva Systems Biology Corp PLRG1 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP33046_P050
This is a rabbit polyclonal antibody against PLRG1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PLRG1. Alternate names: MGC110980, PRL1, PRP46, PRPF46ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54860