Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLSCR3 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15286620UL

 View more versions of this product

Catalog No. NBP15286620


Add to cart

Description

Description

PLSCR3 Polyclonal antibody specifically detects Antigen in Human, Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

PLSCR3
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
PLSCR3
Synthetic peptides corresponding to PLSCR3(phospholipid scramblase 3) The peptide sequence was selected from the middle region of PLSCR3. Peptide sequence GCGTDTNFEVKTRDESRSVGRISKQWGGLVREALTDADDFGLQFPLDLDV.
32 kDa
20 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
Western Blot
Western Blot 1:100-1:2000
Q9NRY6
Ca(2+)-dependent phospholipid scramblase 3, phospholipid scramblase 3, PL scramblase 3
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
57048
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit