Learn More
Cole-Parmer Parker Hannifin Fitting, PFA, Straight, Threaded Plug, 1/4"NPT(M)  DFS Item
Manufacturer:  Cole-Parmer 3132114
Very good chemical resistance. Rated to 200 psi at 77ºF ( 25ºC). Usable from -65 to 190ºF (-53 to 87ºC).
Catalog No. 50-632-009