Learn More
Cole-Parmer CPC (Colder) DrumQuik Pro Drum Insert Plug  DFS Item
Manufacturer:  Cole-Parmer 3160837
The DrumQuik PRO system offers a simplified solution to your chemical dispensing needs. Build your own system with these individual components. A complete dispensing system maintains a closed system, protecting your facility, your operating environment, and your employees from the potential dangers of chemical drips, spills and escaping fumes. The simple connections, rugged construction, and chemical-resistant materials handle a wide variety of applications—including acids and caustics, agricultural liquids, cleaning agents and sterilants, paints and coatings, and food oils and syrups. The base components are a reusable quick-disconnect coupler and a single-use drum insert assembly; both of which are made from environmentally friendly, food-grade polyethylene (PE). The coupler body and plugs mate equally well with all drum inserts listed.
Catalog No. 50-627-536