Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform Signs Plug Lockouts
Hinge style lockouts seal off plug to prevent equipment from being plugged in
Supplier: Accuform Signs KDD230
Specifications
Hinged Plug Lockout | |
Red | |
Plastic | |
Fits cord size up to 1 in. dia., plug size up to 3 x 6.75 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title