Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ Plungers for SiliaSep™ Solid-Load Cartridges

Plungers for 10 and 55 mL solid-load cartridges, designed to allow injections in a dry form for flash chromatography. Improve injections of compounds soluble only in strong solvents or in large volumes.

$692.00 - $937.00

Specifications

For Use With (Application) Injection of compounds soluble only in strong solvents or in large volumes
Type Plunger for SiliaSep™ Solid-load Cartridges
For Use With (Equipment) SiliaSep™ Solid-load Cartridges, SiliaSep™ Flash Cartridges
Product Line SiliaSep™
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
01-184-508
View Documents
SiliCycle™
AUT0060010
10 mL
Pack for $692.00
Add to Cart
 
01-184-509
View Documents
SiliCycle™
AUT0060055
55 mL
Pack for $937.00
Add to Cart
 
Description

Description

The use of solid-load technique (also called dry-load) is known to improve chromatography resolution, especially for compounds soluble only in strong solvents or in large volumes. Silicycle™'s plungers and solid-load cartridges are designed to allow injections in a dry form on SiliaSep Flash Cartridges.

  • For use with SiliaSep solid-load cartridges and flash cartridges
  • Two formats available: 10 and 55 mL
  • Solvent resistant, to allow the use of any mobile phase for the purification
  • Connect the female luer-lock connection fitting to the flash system (solvent inlet)
  • Insert the bottom part of the plunger inside the solid-load cartridge
Specifications

Specifications

Injection of compounds soluble only in strong solvents or in large volumes
Plunger for SiliaSep™ Solid-load Cartridges
SiliaSep™ Solid-load Cartridges, SiliaSep™ Flash Cartridges
SiliaSep™
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit