Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pluronic™ F-127, 0.2 µm filtered (10% Solution in Water) Non-distribution product as customer accommodation.

Pluronic™ F-127, 0.2 µm filtered (10% Solution in Water)

Pluronic F-127 *10% solution in water* is a nonionic, surfactant polyol (molecular weight approximately 12,500 daltons) that has been found to facilitate the solubilization of water-insoluble dyes and other materials in physiological media. Pluronic F-127 is commonly used to help disperse the acetoxymethyl (AM) esters of our ion indicators as well as our cell tracer dyes such as CFDA-SE.
Form Liquid
Content And Storage Store at room temperature.
Reagent Type Pluronic® F-127
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.