Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PlusOne N,N'-Methylene-bisacrylamide Available on GSA/VA Contract for Federal Government customers only.

Available as a dry powder. GE Healthcare PlusOne N,N'-Methylenebisacrylamide is a general cross-linker used with acrylamide for polyacrylamide gels. It is used in IEF, PAGE and Sequencing.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Additional Information General cross-linker used with acrylamide for polyacrylamide gels. Used in IEF, PAGE and Sequencing.
Solubility Soluble in cold water and hot water, slightly soluble in acetone
Boiling Point 185°C
Melting Point >300°C
Color White
pH 5.5 to 7.0