Learn More
Azzota Corp PLX-4032 DFS Item
Manufacturer:  Azzota Corp C1272
PLX-4032 (Vemurafenib) is an orally bioavailable, ATP-competitive, small-molecule inhibitor of B-Raf(V600E) kinase with potential antineoplastic activity. Please request if need more quantities
Catalog No. 50-161-3173